Assay Details
Gene Symbol: APC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171771.1 | NP_001165242.1 | 7 - 8 | 793 | 82 | |
GenBank mRNA | GQ131429.1 | - | 7 - 8 | 793 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
