Assay Details
Gene Symbol: DIO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001324555.2 | NP_001311484.1 | 1 - 2 | 146 | 87 | |
GenBank mRNA | AF401753.1 | - | 1 - 2 | 146 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
