Assay Details
Gene Symbol: MPZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001358929.1 | NP_001345858.1 | 5 - 6 | 705 | 97 | |
NM_001358930.1 | NP_001345859.1 | 5 - 6 | 705 | 97 | ||
GenBank mRNA | DR425851.1 | - | 5 - 6 | 677 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
