Assay Details
Gene Symbol: CBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204642.1 | NP_989973.1 | 2 - 3 | 218 | 101 | |
GenBank mRNA | AB005619.1 | - | 2 - 3 | 218 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
