Assay Details
Gene Symbol: HSD17B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204837.1 | NP_990168.1 | 3 - 4 | 466 | 58 | |
XM_015299593.1 | XP_015155079.1 | 2 - 3 | 767 | 58 | ||
GenBank mRNA | AB002410.1 | - | 3 - 4 | 466 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
