Assay Details
Gene Symbol: RELA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205129.1 | NP_990460.1 | 6 - 7 | 680 | 88 | |
XM_025145278.1 | XP_025001046.1 | 6 - 7 | 698 | 88 | ||
GenBank mRNA | D13721.1 | - | 6 - 7 | 680 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
