Assay Details
Gene Symbol: BMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007988.1 | NP_001007989.1 | 2 - 3 | 211 | 78 | |
XM_025147344.1 | XP_025003112.1 | 11 - 12 | 2245 | 78 | ||
GenBank mRNA | AY616032.1 | - | 2 - 3 | 211 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
