Assay Details
Gene Symbol: PGLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031588.1 | NP_001026759.1 | 1 - 2 | 385 | 94 | |
GenBank mRNA | AJ720303.1 | - | 1 - 2 | 385 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
