Assay Details
Gene Symbol: CIDEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195123.1 | NP_001182052.1 | 4 - 5 | 522 | 150 | |
GenBank mRNA | BX931231.2 | - | 4 - 5 | 688 | 150 | |
HM165259.1 | - | 4 - 5 | 522 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
