Assay Details
Gene Symbol: SDR42E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195157.1 | NP_001182086.1 | 7 - 8 | 935 | 99 | |
XM_015294226.2 | XP_015149712.1 | 10 - 11 | 1317 | 99 | ||
XM_015294227.2 | XP_015149713.1 | 8 - 9 | 1746 | 99 | ||
XM_015294228.2 | XP_015149714.1 | 9 - 10 | 1785 | 99 | ||
XM_025155108.1 | XP_025010876.1 | 10 - 11 | 1592 | 99 | ||
XM_025155109.1 | XP_025010877.1 | 7 - 8 | 1734 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
