Assay Details
Gene Symbol: CCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197144.1 | NP_001184073.1 | 2 - 3 | 402 | 124 | |
XM_015281095.2 | XP_015136581.1 | 2 - 3 | 332 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
