Assay Details
Gene Symbol: MAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199391.1 | NP_001186320.1 | 3 - 4 | 386 | 133 | |
GenBank mRNA | CB018491.1 | - | 3 - 4 | 386 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
