Assay Details
Gene Symbol: SYPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199458.1 | NP_001186387.1 | 2 - 3 | 293 | 102 | |
GenBank mRNA | CN228269.1 | - | 2 - 3 | 254 | 102 | |
CO635562.1 | - | 2 - 3 | 283 | 102 | ||
CO770683.1 | - | 2 - 3 | 331 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
