Assay Details
Gene Symbol: UBE2W | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199481.1 | NP_001186410.1 | 5 - 6 | 483 | 145 | |
XM_015282784.2 | XP_015138270.1 | 5 - 6 | 741 | 145 | ||
XM_015282785.2 | XP_015138271.1 | 5 - 6 | 462 | 145 | ||
GenBank mRNA | CF257907.1 | - | 5 - 6 | 459 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
