Assay Details
Gene Symbol: PHYHIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199504.1 | NP_001186433.1 | 2 - 3 | 437 | 97 | |
XM_015287963.2 | XP_015143449.1 | 2 - 3 | 500 | 97 | ||
XM_025151505.1 | XP_025007273.1 | 2 - 3 | 327 | 97 | ||
GenBank mRNA | DR425110.1 | - | 2 - 3 | 437 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
