Assay Details
Gene Symbol: JAZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199618.1 | NP_001186547.1 | 1 - 2 | 127 | 85 | |
GenBank mRNA | BU453956.1 | - | 1 - 2 | 127 | 85 | |
BX931062.1 | - | 1 - 2 | 133 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
