Assay Details
Gene Symbol: JAZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199618.1 | NP_001186547.1 | 2 - 3 | 198 | 108 | |
XM_015281472.2 | XP_015136958.1 | 2 - 3 | 175 | 108 | ||
GenBank mRNA | BU453956.1 | - | 2 - 3 | 198 | 108 | |
BX931062.1 | - | 2 - 3 | 204 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
