Assay Details
Gene Symbol: FPGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204815.1 | NP_001191744.1 | 2 - 3 | 214 | 122 | |
XM_015290917.2 | XP_015146403.1 | 2 - 3 | 276 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
