Assay Details
Gene Symbol: STON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204865.2 | NP_001191794.2 | 1 - 2 | 1928 | 114 | |
XM_015278009.2 | XP_015133495.1 | 2 - 3 | 2026 | 114 | ||
XM_015278010.2 | XP_015133496.1 | 3 - 4 | 2202 | 114 | ||
XM_015278011.2 | XP_015133497.1 | 2 - 3 | 2117 | 114 | ||
GenBank mRNA | BU460570.1 | - | 1 - 2 | 512 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
