Assay Details
Gene Symbol: AICDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243222.1 | NP_001230151.1 | 1 - 2 | 166 | 92 | |
GenBank mRNA | AJ446140.1 | - | 1 - 2 | 107 | 92 | |
AJ450317.1 | - | 1 - 2 | 167 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001243222.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
