Assay Details
Gene Symbol: AICDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243222.1 | NP_001230151.1 | 3 - 4 | 580 | 82 | |
GenBank mRNA | AJ446140.1 | - | 3 - 4 | 521 | 82 | |
AJ450317.1 | - | 3 - 4 | 581 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
