Assay Details
Gene Symbol: GPR83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256138.1 | NP_001243067.1 | 2 - 3 | 521 | 82 | |
GenBank mRNA | JF795914.1 | - | 2 - 3 | 521 | 82 | |
JF795916.1 | - | 2 - 3 | 504 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
