Assay Details
Gene Symbol: GPR83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256138.1 | NP_001243067.1 | 3 - 4 | 662 | 98 | |
GenBank mRNA | JF795914.1 | - | 3 - 4 | 662 | 98 | |
JF795915.1 | - | 3 - 4 | 676 | 98 | ||
JF795916.1 | - | 3 - 4 | 645 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
