Assay Details
Gene Symbol: KRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256162.1 | NP_001243091.1 | 3 - 4 | 464 | 131 | |
XM_015290265.2 | XP_015145751.1 | 4 - 5 | 597 | 131 | ||
GenBank mRNA | HQ439437.1 | - | 3 - 4 | 464 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
