Assay Details
Gene Symbol: PGRMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271939.1 | NP_001258868.1 | 2 - 3 | 515 | 145 | |
GenBank mRNA | AJ719868.1 | - | 2 - 3 | 499 | 145 | |
AJ720052.1 | - | 2 - 3 | 514 | 145 | ||
CN210800.1 | - | 2 - 3 | 515 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
