Assay Details
Gene Symbol: GLTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277517.1 | NP_001264446.1 | 2 - 3 | 188 | 136 | |
GenBank mRNA | BX931195.1 | - | 2 - 3 | 188 | 136 | |
CR352559.1 | - | 2 - 3 | 192 | 136 | ||
CR406235.1 | - | 2 - 3 | 199 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
