Assay Details
Gene Symbol: GLTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277517.1 | NP_001264446.1 | 3 - 4 | 328 | 114 | |
GenBank mRNA | BX931195.1 | - | 3 - 4 | 328 | 114 | |
CR352559.1 | - | 3 - 4 | 332 | 114 | ||
CR406235.1 | - | 3 - 4 | 339 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
