Assay Details
Gene Symbol: MGST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277533.1 | NP_001264462.1 | 2 - 3 | 430 | 87 | |
NM_001277537.1 | NP_001264466.1 | 2 - 3 | 177 | 87 | ||
GenBank mRNA | BU355153.1 | - | 2 - 3 | 177 | 87 | |
CN236984.1 | - | 2 - 3 | 430 | 87 | ||
CN383177.1 | - | 2 - 3 | 227 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
