Assay Details
Gene Symbol: MGST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277533.1 | NP_001264462.1 | 3 - 4 | 510 | 99 | |
NM_001277537.1 | NP_001264466.1 | 3 - 4 | 257 | 99 | ||
GenBank mRNA | BU355153.1 | - | 3 - 4 | 257 | 99 | |
CN236984.1 | - | 3 - 4 | 510 | 99 | ||
CN383177.1 | - | 3 - 4 | 307 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
