Assay Details
Gene Symbol: HYI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277570.1 | NP_001264499.1 | 1 - 2 | 305 | 57 | |
GenBank mRNA | BU450299.1 | - | 1 - 2 | 299 | 57 | |
BU459205.1 | - | 1 - 2 | 84 | 57 | ||
CO422157.1 | - | 1 - 2 | 351 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
