Assay Details
Gene Symbol: CIB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277648.1 | NP_001264577.1 | 1 - 2 | 79 | 96 | |
NM_001277649.1 | NP_001264578.1 | 1 - 2 | 79 | 96 | ||
XM_015292132.2 | XP_015147618.1 | 1 - 2 | 131 | 96 | ||
GenBank mRNA | BX932884.1 | - | 1 - 2 | 77 | 96 | |
BX935037.1 | - | 1 - 2 | 72 | 96 | ||
BX935068.1 | - | 1 - 2 | 73 | 96 | ||
DR419188.1 | - | 1 - 2 | 152 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
