Assay Details
Gene Symbol: CIB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277648.1 | NP_001264577.1 | 4 - 5 | 362 | 98 | |
NM_001277649.1 | NP_001264578.1 | 4 - 5 | 362 | 98 | ||
NR_102413.1 | 3 - 4 | 98 | ||||
NR_102414.1 | 3 - 4 | 98 | ||||
XM_015292132.2 | XP_015147618.1 | 4 - 5 | 98 | |||
GenBank mRNA | BU373075.1 | - | 3 - 4 | 98 | ||
BX932884.1 | - | 4 - 5 | 360 | 98 | ||
BX935037.1 | - | 4 - 5 | 355 | 98 | ||
BX935068.1 | - | 4 - 5 | 98 | |||
CD216407.1 | - | 3 - 4 | 98 | |||
DR419188.1 | - | 4 - 5 | 435 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
