Assay Details
Gene Symbol: NDUFA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277655.1 | NP_001264584.1 | 2 - 3 | 316 | 89 | |
GenBank mRNA | BU436010.1 | - | 2 - 3 | 241 | 89 | |
BX930200.1 | - | 2 - 3 | 247 | 89 | ||
BX935467.1 | - | 2 - 3 | 242 | 89 | ||
CR353339.1 | - | 2 - 3 | 256 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
