Assay Details
Gene Symbol: ANGPTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277665.1 | NP_001264594.1 | 3 - 4 | 1396 | 123 | |
GenBank mRNA | BU281899.1 | - | 1 - 2 | 124 | 123 | |
CR387194.1 | - | 2 - 3 | 394 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
