Assay Details
Gene Symbol: ESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277791.1 | NP_001264720.1 | 2 - 3 | 255 | 84 | |
NM_001277792.1 | NP_001264721.1 | 2 - 3 | 130 | 84 | ||
NM_001277793.1 | NP_001264722.1 | 2 - 3 | 113 | 84 | ||
XM_025150364.1 | XP_025006132.1 | 2 - 3 | 258 | 84 | ||
GenBank mRNA | BM427320.1 | - | 2 - 3 | 240 | 84 | |
BU340762.1 | - | 2 - 3 | 130 | 84 | ||
BX930968.2 | - | 2 - 3 | 113 | 84 | ||
BX934861.1 | - | 2 - 3 | 115 | 84 | ||
CR385455.1 | - | 2 - 3 | 130 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
