Assay Details
Gene Symbol: MYOZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277827.1 | NP_001264756.1 | 4 - 5 | 534 | 89 | |
XM_015276275.2 | XP_015131761.1 | 3 - 4 | 461 | 89 | ||
GenBank mRNA | BX935788.2 | - | 4 - 5 | 535 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
