Assay Details
Gene Symbol: PTPMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277882.1 | NP_001264811.1 | 1 - 2 | 186 | 102 | |
GenBank mRNA | BU303873.1 | - | 1 - 2 | 172 | 102 | |
BU342228.1 | - | 1 - 2 | 186 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
