Assay Details
Gene Symbol: PTPMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277882.1 | NP_001264811.1 | 2 - 3 | 276 | 66 | |
GenBank mRNA | BU303873.1 | - | 2 - 3 | 262 | 66 | |
BU342228.1 | - | 2 - 3 | 276 | 66 | ||
CR352799.1 | - | 1 - 2 | 98 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
