Assay Details
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278042.1 | NP_001264971.1 | 3 - 4 | 460 | 140 | |
GenBank mRNA | BU250968.1 | - | 3 - 4 | 275 | 140 | |
BX930465.1 | - | 3 - 4 | 276 | 140 | ||
BX930887.2 | - | 3 - 4 | 303 | 140 | ||
BX933231.1 | - | 3 - 4 | 281 | 140 | ||
CO635559.1 | - | 4 - 5 | 572 | 140 | ||
CR352580.1 | - | 3 - 4 | 303 | 140 | ||
CR353677.1 | - | 2 - 3 | 627 | 140 | ||
CR407506.1 | - | 3 - 4 | 307 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
