Assay Details
Gene Symbol: SENP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293211.1 | NP_001280140.1 | 1 | 2220 | 112 | |
XM_015278828.2 | XP_015134314.1 | 2 | 2253 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
