Assay Details
Gene Symbol: CDC42SE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302117.1 | NP_001289046.1 | 1 - 2 | 368 | 99 | |
XM_004949253.3 | XP_004949310.1 | 1 - 2 | 455 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302117.1, XM_004949253.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
