Assay Details
Gene Symbol: CDC42SE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302117.1 | NP_001289046.1 | 6 - 7 | 1029 | 101 | |
XM_004949253.3 | XP_004949310.1 | 5 - 6 | 1043 | 101 | ||
XM_004949254.3 | XP_004949311.1 | 4 - 5 | 662 | 101 | ||
XM_025144760.1 | XP_025000528.1 | 4 - 5 | 600 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
