Assay Details
Gene Symbol: COX7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302146.1 | NP_001289075.1 | 2 - 3 | 213 | 73 | |
GenBank mRNA | BX936177.2 | - | 1 - 2 | 623 | 73 | |
CN225310.1 | - | 2 - 3 | 213 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
