Assay Details
Gene Symbol: CTNNBIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302158.1 | NP_001289087.1 | 1 - 2 | 34 | 102 | |
XM_015296942.2 | XP_015152428.1 | 2 - 3 | 329 | 102 | ||
XM_025142555.1 | XP_024998323.1 | 2 - 3 | 184 | 102 | ||
GenBank mRNA | AM069930.1 | - | 1 - 2 | 34 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302158.1, XM_015296942.2, XM_025142555.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
