Assay Details
Gene Symbol: SMIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302165.1 | NP_001289094.1 | 3 - 4 | 326 | 62 | |
XM_025149694.1 | XP_025005462.1 | 3 - 4 | 226 | 62 | ||
GenBank mRNA | AJ455359.1 | - | 3 - 4 | 327 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
