Assay Details
Gene Symbol: SLC31A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001305656.1 | NP_001292585.1 | 4 - 5 | 472 | 103 | |
NM_001305657.1 | NP_001292586.1 | 4 - 5 | 369 | 103 | ||
NM_001305658.1 | NP_001292587.1 | 4 - 5 | 361 | 103 | ||
NM_001305659.1 | NP_001292588.1 | 3 - 4 | 287 | 103 | ||
NM_001305660.1 | NP_001292589.1 | 3 - 4 | 275 | 103 | ||
XM_015279516.2 | XP_015135002.1 | 3 - 4 | 344 | 103 | ||
XM_025156080.1 | XP_025011848.1 | 3 - 4 | 360 | 103 | ||
XM_025156081.1 | XP_025011849.1 | 4 - 5 | 661 | 103 | ||
XM_025156082.1 | XP_025011850.1 | 4 - 5 | 386 | 103 | ||
XM_025156083.1 | XP_025011851.1 | 3 - 4 | 593 | 103 | ||
XM_025156084.1 | XP_025011852.1 | 3 - 4 | 771 | 103 | ||
GenBank mRNA | BX929472.1 | - | 4 - 5 | 358 | 103 | |
BX929736.2 | - | 3 - 4 | 294 | 103 | ||
BX934786.2 | - | 2 - 3 | 238 | 103 | ||
BX935134.2 | - | 4 - 5 | 471 | 103 | ||
CR406567.1 | - | 3 - 4 | 287 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
