Assay Details
Gene Symbol: DUSP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318409.1 | NP_001305338.1 | 2 - 3 | 995 | 86 | |
GenBank mRNA | AJ851815.1 | - | 2 - 3 | 995 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
