Assay Details
Gene Symbol: ELOVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318410.1 | NP_001305339.1 | 2 - 3 | 237 | 111 | |
GenBank mRNA | HQ621810.1 | - | 2 - 3 | 237 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
