Assay Details
Gene Symbol: OSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168709.1 | NP_001162180.1 | 2 | 923 | 111 | |
XM_015284947.2 | XP_015140433.1 | 3 | 1347 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
