Assay Details
Gene Symbol: IGBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015278343.1 | XP_015133829.1 | 4 - 5 | 838 | 98 | |
XM_015278344.2 | XP_015133830.1 | 4 - 5 | 838 | 98 | ||
XM_015278345.1 | XP_015133831.1 | 4 - 5 | 838 | 98 | ||
GenBank mRNA | BX929486.1 | - | 4 - 5 | 747 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
